siRNA Details
Gene Symbol: AKAP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270546.1 | NP_001257475.1 | 9 | 1362 | ||
NM_006738.5 | NP_006729.4 | 17 | 5560 | |||
NM_007200.4 | NP_009131.2 | 17 | 5548 | |||
GenBank mRNA | AB055890.1 | - | 18 | 5567 | ||
AB209414.1 | - | 6 | 847 | |||
AF126008.1 | - | 9 | 1202 | |||
AF387101.1 | - | 16 | 5354 | |||
AF406992.1 | - | 17 | 5447 | |||
AK309805.1 | - | 9 | 1231 | |||
BC063592.1 | - | 4 | 335 | |||
BC140862.1 | - | 2 | 177 | |||
BC171798.1 | - | 2 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
