siRNA Details
Gene Symbol: Mrrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026422.2 | NP_080698.1 | 6 | 968 | ||
GenBank mRNA | AK010018.1 | - | 6 | 699 | ||
BC019787.1 | - | 6 | 712 | 4167280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
