siRNA Details
Gene Symbol: Med27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290489.1 | NP_001277418.1 | 4, 5 | 450 | ||
NM_026896.5 | NP_081172.2 | 5, 6 | 724 | |||
GenBank mRNA | AK005251.1 | - | 5, 6 | 699 | ||
BC016537.1 | - | 5, 6 | 715 | 3585027 |
