siRNA Details
Gene Symbol: Gm4983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030401.1 | - | 1346 | 5362444 | ||
BC083083.1 | - | 1204 | 6514154 |
Gene Symbol: 2410018L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063063.1 | - | 4 | 1495 |
Gene Symbol: 9030624G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256489.1 | NP_001243418.1 | 4 | 1469 | ||
GenBank mRNA | AK018572.1 | - | 4 | 1447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
