siRNA Details
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081256.1 | NP_001074725.1 | 15 | 4001 | ||
GenBank mRNA | AK159090.1 | - | 2 | 334 | ||
BC031981.1 | - | 1 | 70 | |||
BC038376.1 | - | 5 | 819 | |||
BC060727.1 | - | 8 | 1561 | |||
BC108415.1 | - | 8 | 1405 | |||
BC151084.1 | - | 15 | 3962 | 9087997 | ||
BC172150.1 | - | 14 | 3362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
