siRNA Details
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168412.1 | NP_001161884.1 | 2, 3 | 420 | ||
NM_005724.5 | NP_005715.1 | 3, 4 | 612 | |||
XM_017021857.1 | XP_016877346.1 | 3, 4 | 221 | |||
GenBank mRNA | AF054840.1 | - | 3, 4 | 338 | ||
AF133423.1 | - | 3, 4 | 412 | |||
AK001305.1 | - | 3, 4 | 458 | |||
AK001310.1 | - | 3, 4 | 444 | |||
AK001326.1 | - | 3, 4 | 555 | |||
AK075336.1 | - | 3, 4 | 493 | |||
AK293677.1 | - | 4, 5 | 401 | |||
AK298157.1 | - | 2, 3 | 301 | |||
BC000704.2 | - | 3, 4 | 513 | 3349181 | ||
BC004280.1 | - | 3, 4 | 474 | 3616380 | ||
BC009248.2 | - | 3, 4 | 477 | 2964568 | ||
BC011206.1 | - | 3, 4 | 506 | 3457158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
