siRNA Details
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081256.1 | NP_001074725.1 | 24 | 5985 | ||
GenBank mRNA | AK018027.1 | - | 5 | 1148 | ||
AK033343.1 | - | 10 | 1909 | |||
AK044708.1 | - | 5 | 2983 | |||
AK129281.1 | - | 15 | 3415 | |||
AK159090.1 | - | 11 | 2311 | |||
BC031981.1 | - | 10 | 2054 | |||
BC038376.1 | - | 14 | 2802 | |||
BC060727.1 | - | 17 | 3545 | |||
BC108415.1 | - | 17 | 3389 | |||
BC151084.1 | - | 24 | 5946 | 9087997 | ||
BC172150.1 | - | 23 | 5306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
