siRNA Details
Gene Symbol: Ptcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133735.2 | NP_598496.3 | 9 | 2566 | ||
GenBank mRNA | AK004421.1 | - | 1 | 254 | ||
AK088764.1 | - | 9 | 2485 | |||
AK143877.1 | - | 9 | 2525 | |||
AK150514.1 | - | 9 | 2523 | |||
AK151064.1 | - | 9 | 2534 | |||
AK152077.1 | - | 9 | 2534 | |||
AK153366.1 | - | 9 | 2534 | |||
AK166306.1 | - | 1 | 381 | |||
AK171484.1 | - | 9 | 2521 | |||
AK213622.1 | - | 1 | 211 | |||
AK216509.1 | - | 1 | 90 | |||
AK218479.1 | - | 1 | 292 | |||
BC004766.1 | - | 9 | 2484 | 3156304 | ||
BC083097.1 | - | 9 | 2525 | 30459747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
