siRNA Details
Gene Symbol: Olfr111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005485.2 | NP_001005485.1 | 1 | 812 | ||
GenBank mRNA | BC139116.1 | - | 1 | 812 | 8862138 | |
BC139117.1 | - | 1 | 812 | 8862139 |
Gene Symbol: Olfr110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146328.2 | NP_666440.2 | 2 | 874 | ||
GenBank mRNA | BC132347.1 | - | 1 | 818 | 40130624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
