siRNA Details
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175238.5 | NP_780447.4 | 12 | 1407 | ||
GenBank mRNA | AK088688.1 | - | 1 | 99 | ||
AY428571.2 | - | 12 | 1391 | |||
AY585206.1 | - | 11 | 1400 | |||
JX255737.1 | - | 11 | 1343 |
