siRNA Details
Gene Symbol: Arhgap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037727.2 | NP_001032816.1 | 11 | 3043 | ||
NM_001286610.1 | NP_001273539.1 | 10 | 2843 | |||
NM_175476.4 | NP_780685.2 | 11 | 2714 | |||
GenBank mRNA | AK048450.1 | - | 11 | 2674 | ||
AK137526.1 | - | 11 | 3114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
