siRNA Details
Gene Symbol: Mlxip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177582.3 | NP_808250.1 | 9 | 1951 | ||
GenBank mRNA | AK047475.1 | - | 9 | 1951 | ||
BC137770.1 | - | 9 | 1878 | 8860789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
