siRNA Details
Gene Symbol: Duox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177610.2 | NP_808278.2 | 22 | 3002 | ||
GenBank mRNA | AK079781.1 | - | 2 | 162 | ||
BC172138.1 | - | 23 | 3378 | 9054808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
