siRNA Details
Gene Symbol: Aak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040106.2 | NP_001035195.1 | 21 | 14609 | ||
NM_177762.6 | NP_808430.2 | 20 | 14366 | |||
GenBank mRNA | AK053876.1 | - | 1 | 2007 | ||
BC028270.1 | - | 1 | 2003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
