siRNA Details
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005863.2 | NP_001005863.1 | 2 | 1822 | ||
NM_001286413.1 | NP_001273342.1 | 2 | 1704 | |||
GenBank mRNA | AK143781.1 | - | 2 | 1823 | ||
AY626783.1 | - | 1 | 1644 | |||
BC041777.1 | - | 1 | 487 | 4240274 | ||
BC042206.1 | - | 1 | 487 | 4240274 | ||
BC089009.1 | - | 2 | 1704 | 30543240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
