siRNA Details
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145896.1 | NP_001139368.1 | 16 | 4479 | ||
NM_001145897.1 | NP_001139369.1 | 16 | 4423 | |||
NM_198671.2 | NP_941073.2 | 16 | 4611 | |||
GenBank mRNA | AK033263.1 | - | 16 | 4426 | ||
AK042915.1 | - | 16 | 4429 | |||
AK122216.1 | - | 12 | 3653 | |||
AK154829.1 | - | 16 | 4458 | |||
BC043094.1 | - | 16 | 4611 | 6405464 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006255748, XM_008772659, XM_008772661, XM_008772657, XM_008772662, and others.
Gene Ontology Categories:
