siRNA Details
Gene Symbol: Olfr1034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011872.2 | NP_001011872.2 | 1 | 613 |
Gene Symbol: Olfr1036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207142.2 | NP_997025.1 | 1 | 611 | ||
GenBank mRNA | BC145856.1 | - | 1 | 611 | 40131159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
