siRNA Details
Gene Symbol: Olfr1333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207157.2 | NP_997040.1 | 1 | 920 | ||
GenBank mRNA | BC128000.1 | - | 1 | 924 | 40135841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
