siRNA Details
Gene Symbol: Olfr239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207175.2 | NP_997058.2 | 1 | 148 | ||
GenBank mRNA | BC127988.1 | - | 1 | 152 | 40135639 | |
BC134364.1 | - | 1 | 152 | 40135640 |
Gene Symbol: Gm4461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199062.1 | NP_001185991.1 | 1 | 157 |
Gene Symbol: Olfr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010998.2 | NP_035128.2 | 1 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
