siRNA Details
Gene Symbol: Srbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030133.3 | NP_084409.3 | 18 | 2305 | ||
GenBank mRNA | AK144490.1 | - | 2 | 154 | ||
BC038140.1 | - | 2 | 249 | |||
BC100359.1 | - | 11 | 1169 | |||
BC147429.1 | - | 2 | 182 | 9056482 | ||
BC147430.1 | - | 2 | 182 | 9056479 | ||
BC172066.1 | - | 3 | 289 | 9054352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
