siRNA Details
Gene Symbol: Srbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030133.3 | NP_084409.3 | 19, 20 | 2534 | ||
GenBank mRNA | AK144490.1 | - | 3, 4 | 383 | ||
BC038140.1 | - | 3, 4 | 478 | |||
BC100359.1 | - | 12, 13 | 1398 | |||
BC147429.1 | - | 3, 4 | 411 | 9056482 | ||
BC147430.1 | - | 3, 4 | 411 | 9056479 | ||
BC172066.1 | - | 4, 5 | 518 | 9054352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
