siRNA Details
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242396.1 | NP_001229325.1 | 9 | 4442 | ||
NM_207221.2 | NP_997104.2 | 10 | 4210 | |||
GenBank mRNA | AK082106.1 | - | 1 | 335 | ||
AK173162.1 | - | 9 | 4212 | |||
BC054559.1 | - | 1 | 773 | |||
BC068318.1 | - | 1 | 767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
