siRNA Details
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242396.1 | NP_001229325.1 | 11 | 5618 | ||
NM_207221.2 | NP_997104.2 | 12 | 5383 | |||
GenBank mRNA | AK082106.1 | - | 3 | 1511 | ||
AK173162.1 | - | 11 | 5388 | |||
BC054559.1 | - | 3 | 1949 | |||
BC068318.1 | - | 3 | 1943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
