siRNA Details
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207231.1 | NP_997114.1 | 6 | 885 | ||
GenBank mRNA | AK138099.1 | - | 2 | 1712 | ||
AK154137.1 | - | 6 | 586 | |||
BC065791.1 | - | 6 | 885 | 3824354 |
