siRNA Details
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035247.2 | NP_001030324.2 | 2, 3 | 265 | ||
NM_005659.6 | NP_005650.2 | 2, 3 | 265 | |||
XM_011530361.2 | XP_011528663.1 | 2, 3 | 391 | |||
GenBank mRNA | AF141201.1 | - | 1, 2 | 135 | ||
AJ239058.1 | - | 1, 2 | 135 | |||
AK225877.1 | - | 2, 3 | 222 | |||
AK304734.1 | - | 2, 3 | 235 | |||
AK315998.1 | - | 2, 3 | 216 | |||
AY101594.1 | - | 2, 3 | 192 | |||
BC001049.2 | - | 2, 3 | 212 | 3507963 | ||
BC005087.1 | - | 2, 3 | 213 | 3830774 | ||
CB128364.1 | - | 2, 3 | 265 | |||
CR456607.1 | - | 2, 3 | 149 | |||
DB496066.1 | - | 2, 3 | 249 | |||
U64444.1 | - | 2, 3 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
