siRNA Details
Gene Symbol: 4921511C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076563.1 | - | 1 | 716 | ||
BC125512.1 | - | 1 | 675 | |||
BC132192.1 | - | 1 | 675 |

More Information
Other Species Targeted

