siRNA Details
Gene Symbol: Zfp532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207255.2 | NP_997138.1 | 6 | 4449 | ||
GenBank mRNA | AK163988.1 | - | 7 | 2429 | ||
BC046409.1 | - | 6 | 2126 | |||
BC067032.1 | - | 6 | 4450 | 6841127 | ||
BC094671.1 | - | 8 | 4812 | 30618961 |
