siRNA Details
Gene Symbol: Gm5108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256184.1 | NP_001243113.1 | 2 | 229 | ||
GenBank mRNA | AK076825.1 | - | 2 | 231 | ||
BC120772.1 | - | 2 | 159 | |||
BC120798.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
