siRNA Details
Gene Symbol: Scgb2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207262.2 | NP_997145.2 | 2 | 137 | ||
GenBank mRNA | AY370634.1 | - | 2 | 145 | ||
CK616217.1 | - | 2 | 137 | |||
GU269234.1 | - | 2 | 124 |
