siRNA Details
Gene Symbol: Scgb2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207262.2 | NP_997145.2 | 3 | 281 | ||
GenBank mRNA | AY370634.1 | - | 3 | 289 | ||
CK616217.1 | - | 3 | 281 | |||
GU269234.1 | - | 3 | 268 |
