siRNA Details
Gene Symbol: BC052040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145898.1 | NP_001139370.1 | 11 | 2748 | ||
NM_207264.4 | NP_997147.3 | 11 | 2730 | |||
GenBank mRNA | AK090080.1 | - | 11 | 2673 |
