siRNA Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207270.2 | NP_997153.2 | 9 | 1679 | ||
GenBank mRNA | AK078884.1 | - | 8 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
