siRNA Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207270.2 | NP_997153.2 | 20 | 3227 | ||
GenBank mRNA | AB217856.1 | - | 19 | 2900 | ||
AK078884.1 | - | 19 | 2904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
