siRNA Details
Gene Symbol: D630004N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA863672.1 | - | 1 | 240 | ||
AK085283.1 | - | 1 | 1628 | |||
BC090640.1 | - | 1 | 1831 | |||
BC096037.1 | - | 1 | 1831 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D630004N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA863672.1 | - | 1 | 240 | ||
AK085283.1 | - | 1 | 1628 | |||
BC090640.1 | - | 1 | 1831 | |||
BC096037.1 | - | 1 | 1831 |