siRNA Details
Gene Symbol: Ccdc121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207280.3 | NP_997163.2 | 1 | 1068 | ||
GenBank mRNA | AK078370.1 | - | 1 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
