siRNA Details
Gene Symbol: TRIM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015906.3 | NP_056990.3 | 6 | 1221 | ||
NM_033020.2 | NP_148980.2 | 6 | 1221 | |||
XM_005270936.3 | XP_005270993.1 | 6 | 1142 | |||
XM_005270937.3 | XP_005270994.1 | 6 | 1142 | |||
XM_011541568.2 | XP_011539870.1 | 6 | 1142 | |||
XM_017001454.1 | XP_016856943.1 | 6 | 1142 | |||
GenBank mRNA | AF119043.1 | - | 6 | 1221 | ||
AF220136.1 | - | 6 | 1221 | |||
AF220137.1 | - | 6 | 1221 | |||
AK023434.1 | - | 3 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
