siRNA Details
Gene Symbol: Olfr1118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207632.2 | NP_997515.2 | 1 | 234 | ||
GenBank mRNA | BC120792.1 | - | 1 | 234 | 40129715 |
Gene Symbol: Olfr1116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011734.1 | NP_001011734.1 | 1 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
