siRNA Details
Gene Symbol: Tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009323.2 | NP_033349.2 | 8 | 2779 | ||
GenBank mRNA | AF041822.1 | - | 8 | 2686 | ||
AK132578.1 | - | 8 | 2780 | |||
AK137458.1 | - | 8 | 2576 | |||
AY662679.1 | - | 8 | 2700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
