siRNA Details
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013716.2 | NP_038744.1 | 10 | 1046 | ||
GenBank mRNA | AB001927.1 | - | 10 | 1046 | ||
AK141689.1 | - | 10 | 1113 | |||
AK182641.1 | - | 3 | 242 | |||
AK220230.1 | - | 10 | 1100 | |||
BC021156.1 | - | 10 | 1058 | 4020362 |
