siRNA Details
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013716.2 | NP_038744.1 | 12 | 2056 | ||
GenBank mRNA | AB001927.1 | - | 12 | 2055 | ||
AK141689.1 | - | 11 | 2013 | |||
AK164212.1 | - | 1 | 332 | |||
AK220230.1 | - | 12 | 2109 |
