siRNA Details
Gene Symbol: Clnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013748.3 | NP_038776.3 | 8 | 565 | ||
GenBank mRNA | AB021220.1 | - | 8 | 685 | ||
AF187819.1 | - | 8 | 565 | |||
AK082826.1 | - | 4 | 303 | |||
BC125637.1 | - | 8 | 562 | 40130152 | ||
BC125639.1 | - | 8 | 562 | 40130154 |
