siRNA Details
Gene Symbol: Vmn2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019918.2 | NP_064302.2 | 1 | 210 | ||
GenBank mRNA | AB267725.1 | - | 1 | 210 | ||
AF053986.1 | - | 1 | 210 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019918.2 | NP_064302.2 | 1 | 210 | ||
GenBank mRNA | AB267725.1 | - | 1 | 210 | ||
AF053986.1 | - | 1 | 210 |