siRNA Details
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030730.2 | NP_109655.2 | 13 | 2415 | ||
GenBank mRNA | AJ132389.1 | - | 13 | 2316 | ||
AK083903.1 | - | 13 | 2396 | |||
AK143506.1 | - | 12 | 2031 | |||
AK155151.1 | - | 13 | 2372 | |||
BC133714.1 | - | 13 | 2264 | 40143565 |
