siRNA Details
Gene Symbol: Tssk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032004.1 | NP_114393.1 | 1 | 323 | ||
GenBank mRNA | AF329484.1 | - | 1 | 323 | ||
AK076653.1 | - | 1 | 50 | |||
BC049542.1 | - | 1 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
