siRNA Details
Gene Symbol: ACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005722.3 | NP_005713.1 | 2, 3 | 382 | ||
GenBank mRNA | AF006082.1 | - | 2, 3 | 241 | ||
AK301634.1 | - | 3, 4 | 367 | |||
AK315205.1 | - | 2, 3 | 231 | |||
BC014546.1 | - | 2, 3 | 248 | 4247021 | ||
BC036253.1 | - | 2, 3 | 390 | 5268748 | ||
BI670123.1 | - | 2, 3 | 319 | |||
DC404298.1 | - | 2, 3 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
