siRNA Details
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF054840.1 | - | 6 | 602 | ||
AF133423.1 | - | 6 | 676 | |||
AK001305.1 | - | 6 | 722 | |||
AK001310.1 | - | 6 | 708 | |||
AK001326.1 | - | 6 | 819 | |||
AK027793.1 | - | 3 | 1927 | |||
AK075336.1 | - | 6 | 757 | |||
AK293677.1 | - | 7 | 665 | |||
BC000704.2 | - | 6 | 777 | 3349181 | ||
M69023.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
