siRNA Details
Gene Symbol: Zfp65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145622.2 | NP_663597.1 | 3 | 326 | ||
GenBank mRNA | AK041786.1 | - | 3 | 273 | ||
BB621429.1 | - | 3 | 327 | |||
BC016248.1 | - | 2 | 235 | 4527362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
