siRNA Details
Gene Symbol: Zfp65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145622.2 | NP_663597.1 | 4 | 1971 | ||
GenBank mRNA | AK135182.1 | - | 1 | 476 | ||
BC016248.1 | - | 3 | 1880 | 4527362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
