siRNA Details
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164573.1 | NP_001158045.1 | 26 | 2599 | ||
GenBank mRNA | AF426467.1 | - | 4 | 280 | ||
AK014505.1 | - | 5 | 529 | |||
BC144867.1 | - | 5 | 504 | 9053415 | ||
BC144868.1 | - | 5 | 504 | |||
BC144869.1 | - | 5 | 504 | |||
BC144870.1 | - | 5 | 480 | 9053418 |
