siRNA Details
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113201.2 | NP_001106672.1 | 7 | 904 | ||
NM_001113202.1 | NP_001106673.1 | 7 | 827 | |||
NM_001113203.2 | NP_001106674.2 | 10 | 2957 | |||
NM_001320193.1 | NP_001307122.1 | 7 | 853 | |||
NM_001320194.1 | NP_001307123.1 | 7 | 827 | |||
NM_005594.5 | NP_005585.1 | 7 | 878 | |||
XM_006719412.1 | XP_006719475.1 | 8 | 7175 | |||
XM_006719413.2 | XP_006719476.1 | 8 | 6431 | |||
XM_006719414.1 | XP_006719477.1 | 8 | 6173 | |||
XM_006719418.1 | XP_006719481.1 | 7 | 610 | |||
XM_006719420.3 | XP_006719483.1 | 7 | 866 | |||
XM_011538389.1 | XP_011536691.1 | 8 | 6173 | |||
XM_017019337.1 | XP_016874826.1 | 9 | 2747 | |||
GenBank mRNA | AK223085.1 | - | 7 | 584 | ||
X80909.1 | - | 7 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
